Would you like to log in again?
TransIT-VirusGEN® Transfection Reagents
Supplier: Mirus Bio
TransIT-VirusGEN® VirusGEN® AAV
MIR6740 MIR6745 MIR6720 MIR6755
77147-424EA 39843.75 USD InStock
77147-424 77147-422 77147-420 77147-418
TransIT-VirusGEN® Transfection Reagents
Transfection Reagents
TransIT-VirusGEN® Transfection reagent is designed to enhance delivery of packaging and transfer plasmids to adherent and suspension HEK 293 cell types to increase recombinant Adeno-Associated virus (AAV) and Lentivirus production.
  • Performance – Efficient DNA delivery for production of high-titer viral vectors
  • Scalability – Efficient production from <1 ml to large scale cultures
  • Flexibility – Compatible with different recombinant virus and cell culture systems
  • Animal origin free – Fully synthetic transfection reagent formulation
TransIT-VirusGEN® is ideal for academic research and biopharmaceutical discovery. Titers can be further increased with optimized enhancers included in the VirusGEN® AAV Transfection Kit and VirusGEN® LV Transfection kit.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty