Would you like to log in again?
RightCare 3-Ply Face Masks for General Use
Supplier: Outsource Pharmacy Solutions
RightCare
RCTPFMBX RCTPFMCS
77148-104CS 280 USD InStock
77148-104 77148-102
RightCare 3-Ply Face Masks for General Use
Masks Hygiene Masks
RightCare's 3-ply disposable face masks are a class 1 device for general use PPE. These protective, single-use face masks feature 3 layers of protection that help to filter contaminates from entering or exiting the mask while maintaining smooth breathability.
  • General use face masks are made from non-woven fabric
  • Rectangular design measure 6.5" by 3.5"
  • Adjustable and easy to use
  • Lightweight pleated facemask with shapeable nosepiece contours to any face
  • One size fits all
  • Nonsterile and latex-free
General use face masks are made from non woven fabric which provides increased filtration of fluids in air compared to woven cloth. Three layers of protection filter particles from entering and exiting the mask while maintaining smooth breathability. Great for use at work, school, walking, shopping, or running essential errands.
Soft and comfortable inner layer fabric is made of an absorbent poly non-woven fabric that won't irritate the face while in use. Elastic ear strap loops are strong, rounded for comfort, and slip easily over the ears. The outer hydrophobic nonwoven layer helps to prevent fluids from entering the mask. Middle layer provides high level static filtration, preventing micro dust and contaminants from entering.
Packaging: 50 Masks are packaged 50 per box, 40 boxes per case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty