Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Osteopontin ELISA Kit

Supplier: Antibodies.com LLC
A313559-96
77174-044EA 525 USD InStock
77174-044
Human Osteopontin ELISA Kit
Assays ELISAs
Human Osteopontin ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Osteopontin in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 3 to 48 ng/ml
  • Sensitivity: 0.15 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human Osteopontin ELISA kit (A313559) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Osteopontin in serum, plasma or other biological fluids. An antibody specific for Osteopontin has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Osteopontin present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-Osteopontin antibody, which also binds the Osteopontin present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Osteopontin antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Osteopontin antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Osteopontin captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Osteopontin in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty