Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Rotavirus Antigen ELISA Kit

Supplier: Antibodies.com LLC
A314410-96
77175-734EA 525 USD InStock
77175-734
Human Rotavirus Antigen ELISA Kit
Assays ELISAs
Human Rotavirus Antigen ELISA kit is a 90 minutes Indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro qualitative determination of human Rotavirus Antigen in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human Rotavirus Antigen ELISA kit (A314410) employs the indirect enzyme-linked immunoassay technique for the qualitative detection of human Rotavirus Antigen in serum, plasma, and other biological fluids. The antigen recognised by the Rotavirus Antigen has been pre-coated onto a 96-well microtiter plate. Positive controls, negative controls, and samples are added into the wells and incubated. Antibodies in the sample bind to the pre-coated antigen. Unbound antibody is removed in a wash step. An HRP-conjugated detection antibody is then added and incubated. Unbound HRP-conjugated detection antibody is removed in another wash step. TMB substrate is then added, and color develops. The reaction is terminated by the addition of acidic stop solution and the intensity of the yellow color is measured at O.D. 450 nm in a microplate reader. The O.D. 450 nm of samples is then compared to that of the positive and negative controls to determine the presence or absence of human Rotavirus Antigen.

Ordering information: This is a Ready-To-Use ELISA kit that contains: Pre-coated 96 well microplate, positive control, negative control, HRP-conjugated setection sntibody, sample diluent, substrate solution A, substrate solution B, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty