Would you like to log in again?
Human Anti-Respiratory Syncytial Virus IgG Antibody ELISA Kit
Supplier: Antibodies.com LLC
A314418-96
77175-750EA 525 USD InStock
77175-750
Human Anti-Respiratory Syncytial Virus IgG Antibody ELISA Kit
Assays ELISAs
Human Anti-Respiratory Syncytial Virus IgG Antibody ELISA kit is a 90 minute Indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro qualitative determination of human Anti-Respiratory Syncytial Virus IgG antibody in serum, plasma, and other biological fluids.
  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human anti-Respiratory Syncytial Virus IgG antibody ELISA kit (A314418) employs the indirect enzyme-linked immunoassay technique for the qualitative detection of human anti-Respiratory Syncytial Virus IgG antibody in serum, plasma, and other biological fluids. The antigen recognised by the anti-Respiratory Syncytial Virus IgG antibody has been pre-coated onto a 96-well microtiter plate. Positive controls, negative controls, and samples are added into the wells and incubated. antibodies in the sample bind to the pre-coated antigen. Unbound antibody is removed in a wash step. An HRP-conjugated detection antibody is then added and incubated. Unbound HRP-conjugated detection antibody is removed in another wash step. TMB substrate is then added, and color develops. The reaction is terminated by the addition of acidic stop solution and the intensity of the yellow color is measured at O.D. 450 nm in a microplate reader. The O.D. 450 nm of samples is then compared to that of the positive and negative controls to determine the presence or absence of human anti-Respiratory Syncytial Virus IgG antibody.
Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty