Would you like to log in again?
Lakeland CleanMax® Select Disposable Cleanroom Frocks, SMMS
Supplier: Lakeland Industries
Lakeland CleanMax® Select
SBC191CS-MD SBC191CS-XL SBC191CM-SM SBC191CMB-4X SBC191CM-3X SBC191CM-LG SBC191CM-MD SBC191CMB-LG SBC191CMB-XL SBC191CS-3X SBC191CMB-MD SBC191CM-XL SBC191CS-LG SBC191CS-2X SBC191CS-4X SBC191CS-SM SBC191CMB-5X SBC191CS-5X SBC191CMB-3X SBC191CM-2X SBC191CMB-2X SBC191CM-5X SBC191CM-4X SBC191CMB-SM
77188-308CS 216 USD InStock
77188-308 77188-318 77188-306 77188-328 77188-316 77188-326 77188-304 77188-336 77188-314 77188-302 77188-324 77188-312 77188-334 77188-322 77188-300 77188-332 77188-310 77188-320 77188-298 77188-330 77188-296 77188-294 77188-292 77188-290
Lakeland CleanMax® Select Disposable Cleanroom Frocks, SMMS
Disposable Garments
The Lakeland CleanMax® Select frock is designed with breathable SMMS fabric that is ideal for ISO Class 5 to 8 cleanrooms.
  • Breathable SMMS fabric
  • Non-sterile and sterile options available
  • Tunnelized, latex-free elastic
  • Low linting
  • Elastic wrists
  • Thumb loops
CleanMax® Select SMMS is available in sterile and non-sterile configurations. Clean-manufactured in Lakeland owned and operated certified cleanrooms, under strict process and environmental controls CleanMax® Select meets IEST particle category cleanliness levels. CleanMax® Select SMMS sterile products are validated to an SAL 10⁻⁶ with sterile traceability down to the individual garment. Resistant to microbial, viral, and blood borne contaminants. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops, and bound seams provide a superior barrier for the wearer. Designed to prevent breakthrough and strikethrough, let Lakeland clean-manufactured CleanMax® Select be the cornerstone of contamination control strategy. Frocks are constructed with a four-layer SMMS cloth-like fabric. Mandarin collar, zipper with placket, elastic wrist with thumb loops.
Caution: Do not use a box cutter to open box. For storage, keep dry and avoid direct sunlight.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty