Would you like to log in again?
Lakeland CleanMax® Select Disposable Cleanroom Coveralls with Hood, Non Sterile, SMMS, Elastic Wrists/Ankles
Supplier: Lakeland Industries
Lakeland CleanMax® Select
SBC428CS-4X SBC428CMB-4X SBC428CMB-SM SBC428CS-LG SBC428CM-3X SBC428CMB-3X SBC428CS-3X SBC428CM-MD SBC428CMB-MD SBC428CS-SM SBC428CS-5X SBC428CMB-LG SBC428CS-MD SBC428CM-SM SBC428CS-XL SBC428CM-4X SBC428CM-XL SBC428CMB-2X SBC428CM-2X SBC428CS-2X SBC428CM-5X SBC428CM-LG SBC428CMB-5X SBC428CMB-XL
77188-438CS 296.77 USD InStock
77188-438 77188-448 77188-436 77188-458 77188-446 77188-468 77188-478 77188-434 77188-456 77188-444 77188-466 77188-454 77188-476 77188-464 77188-442 77188-474 77188-452 77188-462 77188-440 77188-472 77188-450 77188-460 77188-470 77188-480
Lakeland CleanMax® Select Disposable Cleanroom Coveralls with Hood, Non Sterile, SMMS, Elastic Wrists/Ankles
Disposable Garments
The Lakeland CleanMax® Select coverall with hood and elastic wrists and ankles is designed with breathable SMMS fabric that is ideal for ISO Class 5 to 8 cleanrooms.
  • Breathable SMMS fabric
  • Non-sterile and sterile options available
  • Tunnelized, latex-free elastic
  • Low linting
  • Attached hood
  • Elastic wrists and ankles
  • Thumb loops
CleanMax® Select SMMS is available in sterile and non-sterile configurations. Clean-manufactured in Lakeland owned and operated certified cleanrooms, under strict process and environmental controls CleanMax® Select meets IEST particle category cleanliness levels. CleanMax® Select SMMS sterile products are validated to an SAL 10⁻⁶ with sterile traceability down to the individual garment. Resistant to microbial, viral, and blood borne contaminants. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops, and bound seams provide a superior barrier for the wearer. Designed to prevent breakthrough and strikethrough, let Lakeland clean-manufactured CleanMax® Select be the cornerstone of contamination control strategy. The coveralls are constructed with a four-layer SMMS cloth-like fabric. Attached hood, zipper with placket, elastic ankles and wrist with thumb loops.
Caution: Do not use a box cutter to open box. For storage, keep dry and avoid direct sunlight.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty