Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,1,1-Trichloro-2-methyl-2-propanol hemihydrate 98%

Supplier: Ambeed
Warning
Synonyms: Chlorbutanol hemihydrateChlorobutanol hydrous1,1,1-Trichloro-2-methyl-2-propanol Hemihydrate

A354411-10G
77195-244EA 6 USD InStock
77195-244
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate 98%
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate
Caution: for Research Use Only


Formula: C₈H₁₆Cl₆O₃
MW: 372,93 g/mol
MDL Number: MFCD02179352
CAS Number: 6001-64-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty