Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Staticide® 8117 Premium Maroon Wrist Strap, 4 mm

Supplier: ACL Staticide
Staticide®
8117
77200-596EA 25.5 USD InStock
77200-596
Staticide® 8117 Premium Maroon Wrist Strap, 4 mm
Grounders Wrist Grounders
This adjustable premium maroon-colored wrist strap provides proper grounding for ESD applications.

  • Recommended for all-day wear
  • 2mm band thickness
  • Elastic crochet fabric lined with 16 rows of silver yarn
  • Flat buckle back
  • 4 mm to banana, single wire

Conductive fibers are interwoven for maximum skin contact and minimum skin irritation. Lightweight design allows for all-day wear in electrostatic protected areas and manufacturing. Designed for use with the 8107 wrist strap cord.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty