Would you like to log in again?
Staticide® 8124 Disposable Heel Strap
Supplier: ACL Staticide
Staticide®
8124
77200-602PK 39.95 USD InStock
77200-602
Staticide® 8124 Disposable Heel Strap
Grounders Heel and Toe Grounders
The easy to use ACL staticide disposable heel strap is an economical solution for plant visitors and other temporary workers who enter electrostatic protected areas.
  • 99% polyester, 1% conductive yarn
  • Resistance: 10⁵ ohms
These disposable heel straps are packaged 100 strips per bag.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty