Would you like to log in again?
Staticide® 8150 Economy ESD Wrist Strap Kit, 4 mm
Supplier: ACL Staticide
Staticide®
8150
77200-604KT 10.95 USD InStock
77200-604
Staticide® 8150 Economy ESD Wrist Strap Kit, 4 mm
Grounders Wrist Grounders
Adjustable economy wrist strap kit offers comfortable and secure personal grounding for occasional use.
  • Includes dark blue wrist strap, cord, and alligator clip
  • Recommended for occasional wear
  • Elastic crochet fabric linded with 5 rows of double-stitched silver yarn
  • 1.5 mm band thickness
Once connected to ground, the wrist strap provides a safe path for eliminating static charges while working on electronics.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty