Would you like to log in again?
Staticide® IPA Aerosol
Supplier: ACL Staticide
Staticide®
8625
77200-606CS 67.32 USD InStock
77200-606
Staticide® IPA Aerosol
Cleaning Agents Detergents and Surface Cleansers
Ideal choice for general cleaning applications on a variety of substrates where removing contaminants are of concern.
  • Flammable
  • 99.8% pure isopropyl alcohol
  • Made in the USA
Extra-strength IPA aerosol is safe for use on plastics. Evaporates quickly, dries fast, and leaves no residue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty