Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Staticide® Conductive Trays

Supplier: ACL Staticide
Staticide®
972618 972015
77200-612EA 53.25 USD InStock
77200-612 77200-610
Staticide® Conductive Trays
Trays/Dishes/Pots Laboratory Trays/Dishes
Staticide® conductive trays are designed to protect against ESD when used for assembling, soldering, or carrying static-sensitive components.

  • Shiny smooth finish
  • Temperature range: 5 to 107 °C (40 to 225 °F)
  • Conductivity: less than 10⁶ ohms RTT, RTG, Vol R
  • Compatible with cart transport systems
  • Resistant to abrasion and chemicals

Certifications: RoHS3, REACH compliant.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty