Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Staticide® Dualmat™ ESD Mats, Bulk Rolls, Blue

Supplier: ACL Staticide

Staticide®
7275RBR3040 7275RBR2440
77200-656EA 1100 USD InStock
77200-656 77200-654
Staticide® Dualmat™ ESD Mats, Bulk Rolls, Blue
Surface Protectors Bench Protectors
The Dualmat™ was developed for companies adhering to electrostatic control and lead-free requirements.

  • Top side resistance measures 10⁶ to 10⁸ ohms
  • Bottom side resistance measures less than 5×10⁵ ohms
  • Embossed surface
  • Rolls available in lengths of 40' and widths of 24", 30" and 36"
  • Custom cut sizes available
  • Mat thickness is 0.15 cm (0.06")

The durable, 2-layer rubber material is inherently dissipative on the top side and conductive on the bottom side. These mats are ideal for electronics manufacturing. Includes 4 oz. mat cleaner.

Certifications: Meets ANSI/ESDA S20.20.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty