Would you like to log in again?
Staticide® ACL Microdenier Wipes
Supplier: ACL Staticide
Staticide®
8299MD
77200-698CS 383.7 USD InStock
77200-698
Staticide® ACL Microdenier Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
A unique fabric meets the highest standard for critical cleaning.
  • Super soft material, does not scratch
  • Ultra-low particle and fiber generation
  • Suitable for ISO Class 4 – 5 (Class 10-100) environments
  • Class 10 laundered and packaged
Made from a 95% polyester/5% nylon blend, these wipes are designed to attract and hold contaminants within a wedge-like structure.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty