Would you like to log in again?
Staticide® ACL Microfiber Wipes
Supplier: ACL Staticide
Staticide®
8409MF 8404MF
77200-702CS 329.75 USD InStock
77200-702 77200-704
Staticide® ACL Microfiber Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
The ACL Staticide microfiber wipe for critical task cleaning is made from hydro-entangled split microfiber. This material provides excellent pick-up and removal of contaminants, whether used wet or dry.
  • 80% polyester/20% nylon hydro-entangled blend
  • Binder-free, no chemical residue
  • Ultra-low particle and fiber release
  • Knife-cut edge
Offering excellent durability, strength, and liquid absorbancy, the Microfiber wipes are suitable for use in ISO Class 5 (Class 100) environments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty