Would you like to log in again?
Staticide® ESD Resealable Metal-In Shielding Bags
Supplier: ACL Staticide
Staticide®
MIZ1012 MIZ46 MIZ35 MIZ610 MIZ23 MIZ58 MIZ68
77200-768CS 159.75 USD InStock
77200-768 77200-778 77200-776 77200-774 77200-772 77200-780 77200-770
Staticide® ESD Resealable Metal-In Shielding Bags
Bags Storage Bags
Resealable static shielding bags provide protection for PCB components from static and ESD.
  • Metalized within layers
  • 2.8 mil thickness ±10%
  • Static shielding <15nJ
  • Surface resistance (outside and inside) <10¹¹ ohms
  • ESD logo on every bag
  • Traceable date codes
Three layer construction features static dissipative polyethylene (inner), aluminum (middle), and static dissipative polyester (outer).
Certifications: Meets ANSI/ESD S541. RoHS compliant.
Packaging: Bags are packaged 100 per pack, 5 packs to a case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty