Would you like to log in again?
VWR Opti-Swab® Liquid Amies Collection and Transport Systems
Supplier: Avantor
VWR Opti-Swab®
LA-116VWR LA-117VWR LA-106VWR
77200-796CS 406.38 USD InStock
77200-796 77200-794 77200-792
VWR Opti-Swab® Liquid Amies Collection and Transport Systems
Cell Culture Media
The Opti-Swab® liquid amies transport system is ideal for the collection and transport of aerobic, anaerobic, and fastidious bacteria. It is also suitable for use in bacterial culture, gram staining, rapid tests, automation, and molecular tests when validated by test manufacturers and/or laboratory.
  • Ready-to-use liquid amies collection and transport system
  • For the collection and transport of aerobic, anaerobic, and fastidious bacteria
  • Suitable for use in bacterial culture, gram staining, rapid tests, automation and molecular tests when validated by test manufacturers and/or laboratory
  • Advanced kit design is easy to use and reliable
  • Self-centering cap is leak-proof and facilitates easy swab removal and disposal
  • Soft tip for patient comfort
  • Premium medical grade plastic components to improve product shelf life
  • Shelf life: 18 months
  • For In Vitro diagnostic use
  • Made in USA
High-performance swab tips are available in three formats. The HydraFlock swab (3317-H) with 100 mm molded breakpoint is designed to accommodate nasopharyngeal (NP) sampling sites. The HydraFlock swab (3316-H) with molded breakpoint is designed to accommodate eye, ear, urogenital, and pediatric sampling sites. The HydraFlock swab (3506-H) with molded breakpoint is designed to accommodate nose, throat, vaginal, rectal, and wound sampling sites.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty