Would you like to log in again?
VWR UniTranz-RT® Transport System
Supplier: Avantor
VWR UniTranz-RT®
UT-317VWR
77200-798CS 1193.27 USD InStock
77200-798
VWR UniTranz-RT® Transport System
Cell Culture Media
The UniTranz-RT® universal transport is ideal for the collection and preservation of viruses including, chlamydia, mycoplasma, and urea plasma. It is also suitable for use with antigen detection, PCR, and rapid tests when validated by test manufacturers and/or laboratory.
  • Premium medical grade plastic components improve product shelf life
  • PurFlock Ultra swab: 25-3317-U
  • Shelf life: 18 months
  • For In Vitro diagnostic use
  • Made in USA
All kits include vial, and individually wrapped sterile swabs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty