Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human FAM83H ELISA Kit

Supplier: Antibodies.com LLC
A310427-96
77201-720EA 525 USD InStock
77201-720
Human FAM83H ELISA Kit
Assays ELISAs
Human FAM83H ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human FAM83H in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 75 - 1200 ng/L
  • Sensitivity: 4.34 ng/L
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Human FAM83H ELISA kit (A310427) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human FAM83H in serum, plasma or other biological fluids. An antibody specific for FAM83H has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the FAM83H present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-FAM83H antibody, which also binds the FAM83H present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-FAM83H antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-FAM83H antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of FAM83H captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of FAM83H in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty