Would you like to log in again?
Human CD32 ELISA Kit
Supplier: Antibodies.com LLC
A312148-96
77202-616EA 525 USD InStock
77202-616
Human CD32 ELISA Kit
Assays ELISAs
Human CD32 ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human CD32 in serum, plasma, and other biological fluids.
  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human CD32 ELISA kit (A312148) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human CD32 in serum, plasma or other biological fluids. An antibody specific for CD32 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the CD32 present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-CD32 antibody, which also binds the CD32 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-CD32 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-CD32 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of CD32 captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of CD32 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, positive control, negative control, HRP-conjugated detection antibody, sample diluent, substrate solution A, substrate solution B, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty