Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EZ FLO Hypodermic Needles

Supplier: Air-Tite
EZ FLO
NEZ22100 NEZ23100 NEZ22300 NEZ22200 NEZ18112 NEZ27112 NEZ20112 NEZ22112 NEZ23112 NEZ25200 NEZ25112 NEZ20100 NEZ21112 NEZ27114 NEZ27100 NEZ25100 NEZ21200 NEZ22400 NEZ21100 NEZ18100 NEZ19112 NEZ22034 NEZ18200 NEZ30100 NEZ25058 NEZ27200 NEZ19100 NEZ18034 NEZ23034 NEZ30012
77232-948BX 10.14 USD InStock
77232-948 77232-950 77232-972 77232-994 77232-974 77232-952 77232-996 77232-976 77232-954 77232-956 77232-978 77232-990 77232-970 77232-992 77232-958 77232-938 77232-962 77232-984 77232-940 77232-942 77232-964 77232-986 77232-966 77232-988 77232-944 77232-968 77232-946 77232-980 77232-982 77232-960
EZ FLO Hypodermic Needles
Syringe Needles
The EZ FLO line of general purpose hypodermic needles by Air-Tite Products.

  • High quality
  • Low cost
  • Broad range
  • Standard bevel
  • Thin wall 18 to 25 g
  • Regular wall 27 and 30 g

Air-Tite Products' EZ FLO line of hypodermic needles offer high quality and a broad selection at an affordable price. Ranging from 18 to 30 gauge, these needles have a standard bevel with thin wall from 18 to 25 g and regular wall in 27 and 30 g to combine comfort with durability.

Made of stainless steel with a polypropylene hub. Sterilized by ethylene oxide.

Certifications: Complies with ISO 13485 and ISO 7864.

Packaging: Each needle is sterile, individually blister packaged in a box of 100.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty