Would you like to log in again?
VWR® Bagged Examination Gloves, Powder-Free, Nitrile
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
DCF65NBWH-0-L DCF65NBWH-0-S DCF65NBBL-0-XL DCF65NBBL-0-S DCF65NBWH-0-M DCF65NBBL-0-M DCF65NBBL-0-L DCF65NBWH-0-XL
77257-068CS 220 USD InStock
77257-068 77257-058 77257-060 77257-070 77257-062 77257-072 77257-064 77257-066
VWR® Bagged Examination Gloves, Powder-Free, Nitrile
Gloves
VWR® bagged examination gloves provide quality and comfort. Perfect for cleanroom, laboratory, and ESD environments.
  • Bagged gloves for cleanroom compatibility
  • Cleanroom class 10,000 (DI washed)
  • Static control
  • CAFE RGP: 10⁹ ohms as per ANSI/ESD SP15.1
  • Surface resistance: 10¹¹ Ω
  • Volume resistance: 10⁹ Ω
VWR® bagged examination gloves are powder-free nitrile, cleanroom compatable gloves that offer static control and provide a high level of quality and comfort. These gloves are available in a blue or white finish.
Certifications: Meets ANSI/ESD SP15.1.
Packaging: Gloves are packaged 100 per double bag, 10 bags per case (1000 gloves per case).
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty