Would you like to log in again?
LocHook® Double Rod Zinc Plated Steel Pegboard Hooks for LocBoard®
Supplier: Triton Products
52419 52628 52319 52213 52119 52113 52313
77269-244PK 22.93 USD InStock
77269-244 77269-256 77269-254 77269-248 77269-246 77269-252 77269-250
LocHook® Double Rod Zinc Plated Steel Pegboard Hooks for LocBoard®
Wall Hooks
The patented LocHook® provides the ultimate building block system for secure organization and storage.
  • Ideal for storing hammers, sledge hammers, large hand tools and power tools when used with other LocHooks
  • The 2 and 4-point contact locking mechanism offers industrial strength holding capability
  • Hooks mount vertically and horizontally in 4 distinct positions allowing the use of multiple hooks for large, heavy and odd-shaped items
  • Fixtures install from the front, so no need to remove the board for installation or repositioning of LocHooks® causing no damage to the LocBoard®
  • Compatible with all LocBoard® sizes and other brands of steel tool boards with square holes spaced 1 to ¹/₂" O.C.
Having secure, commercial grade, easily accessible and versatile storage and organizational solutions is an issue that every business encounters. LocHook®(s) and LocBoard® present an ideal steel pegboard tool storage solution for companies that support 5S and Lean Manufacturing initiatives. They provide the most heavy-duty pegboard and hook system and are ideal for dependable storage for factory floor, shop, home and garage. The LocHook is an innovative pegboard hook which can be mounted in any direction and is rugged enough to support just about anything. Together, the LocBoard and LocHook will provide the perfect steel pegboard solution. Imagine a clean looking commercial grade pegboard with the strongest hooks available to hang your items within grasp at all times. That is the power of the steel pegboard system from Triton Products.
Ordering information: Limited lifetime warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty