Would you like to log in again?
Stainless Steel LocHook® Single Rod Pegboard Hooks for Stainless Steel LocBoard®
Supplier: Triton Products
61113 61629 61323
77269-292PK 14.75 USD InStock
77269-292 77269-290 77269-294
Stainless Steel LocHook® Single Rod Pegboard Hooks for Stainless Steel LocBoard®
Wall Hooks
The stainless steel LocHook® is ideal for food, medical, healthcare, restaurant, marine and commercial applications as well as any clean room or harsh environment that must meet stringent requirements.
  • Single rod hooks are offered with multiple degree bends to accommodate any size tool assuring items will not slide off
  • Easily repositioned to accommodate changing storage needs with no damage to the stainless steel LocBoard
  • Won't rust or corrode in any environment, even ones with extreme temperatures
  • Use multiple hooks for large, heavy and odd-shaped items
Having clean, easily accessible and versatile storage and organizational solutions is an issue that every business encounters. The food and healthcare industries must meet stringent requirements and The stainless steel pegboard storage from Triton products provides the ideal answer. Two innovative best-selling products, the LocBoard and the LocHook, are now available in stainless steel. They provide a commercial grade heavy-duty pegboard and hooks system. The LocHook is an innovative pegboard hook which can be mounted in any direction and is rugged enough to support just about anything. Together, the LocBoard and LocHook will provide the perfect stainless steel pegboard solution. Imagine a clean, well-kept kitchen with every pan, ladle, knife and utensil conveniently hanging and within grasp at all times. Imagine every medical supply in a hospital being easily accessible at a moment's notice. That is the power of the stainless steel pegboard system from Triton products.
Ordering information: Limited lifetime warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty