Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Pegboard Hook, 2¹/₄" Curved 2" Int.Ø Stainless Steel, for ¹/₈" and ¹/₄" Pegboard
Supplier: Triton Products
85200
77269-390PK 10.6 USD InStock
77269-390
Pegboard Hook, 2¹/₄" Curved 2" Int.Ø Stainless Steel, for ¹/₈" and ¹/₄" Pegboard
Wall Hooks
The stainless steel DuraHook® is ideal for food, medical, healthcare, restaurant, marine and commercial applications as well as any clean room or harsh environment that must meet stringent requirements.
  • Use in pairs to hold power tools including drills, caulk guns and many other long items
  • Innovative double locking system locks the hook in place with a locking tab that mounts to the pegboard from the front
  • Won't rust or corrode in any environment, even ones with extreme temperatures
  • Use on any ¹/₈" or ¹/₄" pegboard (excluding steel)
  • Hooks mount vertically and horizontally in four distinct positions allowing the use of multiple hooks for large, heavy and odd-shaped items
  • Fixtures install from the front, so no need to remove the board for installation or repositioning of stainless steel DuraHooks
Having clean, easily accessible and versatile storage and organizational solutions is an issue that every business encounters. The food and healthcare industries must meet stringent requirements and The stainless steel pegboard storage from Triton products provides the ideal answer. Our innovative DuraHook is now available in stainless steel. The stainless steel DuraHook is an innovative pegboard hook which can be mounted in any direction and is rugged enough to support just about anything. Imagine a clean, well-kept kitchen with every pan, ladle, knife and utensil conveniently hanging and within grasp at all times. Imagine every medical supply in a hospital being easily accessible at a moment's notice. That is the power of The stainless steel DuraHook from Triton products.
Ordering information: Limited lifetime warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty