Would you like to log in again?
DuraBoard® Pegboards, Black ABS Textured
Supplier: Triton Products
DuraBoard®
DB-96BLK DB-4BK DB-2BK
77269-622EA 414.51 USD InStock
77269-622 77269-602 77269-614
DuraBoard® Pegboards, Black ABS Textured
Furniture Backsplashes, Wallracks, Wallpanels
DuraBoard® is the most durable, weather and impact resistant pegboard available. It is eight times stronger than conventional pegboard.
  • Hole diameter is ¹/₄" with 1" O.C spacing
  • DuraBoard will never fade, dry out, crack or peel over time so it maintains holding capability over many years of use
  • Offers a professional looking pegboard system that can be cleaned without harming the board
  • Board surface is ideal for creating silhouettes of tools to be stored and managed allowing for easy tool identification
  • Use Triton 70016 mounting spacer kit to mount DuraBoard and eliminate the expense and hassle of wood framing
The commercial-grade DuraBoard® ¹/₄" textured ABS pegboard has superior holding strength with eight times that of traditional pegboard. One side has a textured finish to keep it looking new and to prevent scratching. It is the most durable, weather and impact resistant pegboard available. This pegboard will stand up to any harsh temperatures and environments and can be cleaned or washed when necessary. DuraBoard® heavy-duty pegboard is ideal for use in a clean room, factory floor production area, maintenance and repair area as well as sheds and garages. Use DuraHook® double-locking pegboard hooks to create the most secure and reliable pegboard storage system.
Ordering information: Limited lifetime warranty.
Delivery information: Mounting hardware not included.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty