Would you like to log in again?
Two Pegboards, LocHook® and Hanging Bin Assortment, 18-Gauge Steel Square Hole
Supplier: Triton Products
LB18-BKKIT LB18-CK LB18-BKIT LB18-GKIT
77269-650EA 268.39 USD InStock
77269-650 77269-654 77269-648 77269-646
Two Pegboards, LocHook® and Hanging Bin Assortment, 18-Gauge Steel Square Hole
Furniture Backsplashes, Wallracks, Wallpanels
LocBoard® is made from 18 gauge steel and offers the ultimate holding capacity for the most demanding tool and storage needs. It is 12 times stronger than conventional pegboard.
  • Ideal for heavy, oversized and odd shaped professional power tools, hand, maintenance tools and more
  • Two boards can be mounted vertically or horizontally together or positioned on multiple wall areas as required
  • Use with LocHook® 50000 Series 100% heavy-duty steel 2 and 4-point contact locking hooks
  • All hooks lock securely on the board and are easily repositioned
  • Mounting hardware is hidden from sight - presenting a clean, attractive and durable appearance
  • 350 lbs. load rating per board
LocBoard® heavy-duty scratch resistant epoxy coated steel pegboard with square holes offers ultimate holding capacity for demanding storage needs. LocBoard® kit is ideal for use in clean room, factory floor production, maintenance, repair areas, home and garage. Industrial strength LocHook®s create the most reliable pegboard system available. Demanding tool storage requirements for power tools, large and odd shaped tools are easily met using LocBoard steel pegboards.
Ordering information: Limited lifetime warranty.
Delivery information: Contents include (2) 18×36" LocBoards; 28 hooks: (1) Multi-prong tool holder; (1) multi-ring tool holder; (2) 4" Single rod 30° bend; (2) 6" single rod 90° bend; (2) 5¾" Double rod 80° bend; (3) 1" Single rod 30° bend; (3) 3" Single rod 30° bend; (3) 3" double rod 90° bend; (3) 2-¼" Curved hook; (2) Extended spring clip; (2) Standard spring clips; (2) closed hammer/pliers holder; (2) BinClips; (2) black plastic hanging bins; mounting hardware included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty