Would you like to log in again?
Two Pegboards, 18-Gauge Steel Square Hole with Mounting Hardware
Supplier: Triton Products
LB1-B LB2-B LB2-G LB2-W LB2-R LB1-G LB1-W
77269-662PK 192 USD InStock
77269-662 77269-696 77269-682 77269-688 77269-666 77269-678 77269-676
Two Pegboards, 18-Gauge Steel Square Hole with Mounting Hardware
Furniture Backsplashes, Wallracks, Wallpanels
LocBoard® is made from 18 gauge steel and offers the ultimate holding capacity for the most demanding tool and storage needs. It is 12 times stronger than conventional pegboard.
  • Ideal for heavy, oversized and odd shaped professional power tools, hand, maintenance tools and more
  • Two boards can be mounted vertically or horizontally together or positioned on multiple wall areas as required
  • Use with LocHook® 50000 series 100% heavy-duty steel two and four point contact locking hooks
  • All hooks lock securely on the board and are easily repositioned
  • Mounting hardware is hidden from sight - presenting a clean, attractive and durable appearance
LocBoard® heavy-duty Epoxy coated steel pegboards with square holes offer the ultimate holding capacity for the most demanding tool and storage needs. The Epoxy coating makes them scratch resistant. LocBoard® heavy-duty steel pegboard is ideal for use in a clean room, factory floor production area, maintenance, repair areas, home and garage. Use only LocHook® 50000 series industrial strength hooks and steel shelf with patented two and four point contact locking mechanisms to create the most secure and reliable pegboard system for commercial and home tool storage and other work items. The most demanding tool storage requirements for power tools, large and odd shaped tools are easily met using the LocBoard® steel pegboards.
Ordering information: Limited lifetime warranty.
Delivery information: Mounting hardware included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty