Would you like to log in again?
Two Pegboards, 304 Stainless Steel Square Hole, LocHook® Assortment
Supplier: Triton Products
LB2-SKIT
77269-694EA 835.39 USD InStock
77269-694
Two Pegboards, 304 Stainless Steel Square Hole, LocHook® Assortment
Furniture Backsplashes, Wallracks, Wallpanels
LocBoard® is made from 18 gauge steel and offers the ultimate holding capacity for the most demanding tool and storage needs. It is 12 times stronger than conventional pegboard.
  • Ideal for heavy, oversized and odd shaped professional power tools, hand, maintenance tools and more
  • Two boards can be mounted vertically or horizontally together or positioned on multiple wall areas as required
  • Use with LocHook® 50000 Series 100% heavy-duty steel 2 and 4-point contact locking hooks
  • All hooks lock securely on the board and are easily repositioned
  • Mounting hardware is hidden from sight - presenting a clean, attractive and durable appearance
  • 400 lbs. load rating per board
Stainless Steel LocBoard® Kits offer an ultimate pegboard system for clean rooms to harsh environments. Made with 304 brushed stainless steel, LocBoard® heavy-duty pegboards with square holes have more holding capacity for demanding tool and parts storage. Stainless Steel LocHook® 60000 industrial hooks create the most reliable pegboard system. The most dependable pegboard for professional use.
Ordering information: Limited lifetime warranty.
Delivery information: LB2S-Kit includes (2) 24×42¹/₂" Stainless steel LocBoard®; 45 hooks: (12) 1" stainless steelsingle rod 30° bend; (9) 3" stainless steel single rod 30° bend; (3) 6" stainless steel single rod 90° bend; (3) 4" stainless steel double rod 90° bend; (9) stainless steel extended spring clip; (3) stainless steel standard spring clips; (3) 2¹/₄" stainless steel curved hook; (1) stainless steel multi-prong tool holder; (2) stainless steel multi-ring tool holder, mounting hardware.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty