Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pegboards, Heavy-Duty High Density Fiberboard

Supplier: Triton Products
TPB-2BK TPB-4BK TPB-2W TPB-4W TPB-2N
77269-772PK 170.71 USD InStock
77269-772 77269-762 77269-770 77269-760 77269-758
Pegboards, Heavy-Duty High Density Fiberboard
Furniture Backsplashes, Wallracks, Wallpanels
These pegboards are made of high-grade HDF to ensure durability and are four times stronger than conventional pegboard.

  • Hole diameter is ¹/₄" with 1" O.C spacing
  • Superior holding capacity when compared to standard pegboard
  • Abrasion, humidity and temperature resistant
  • Can be cut (using an 80-tooth or higher saw blade), drilled, or routed without cracking and fraying the board
  • Modulus of Rupture (MOR) rating of 5000 psi
  • Recommended to be used with our DuraHook® 70000 series locking pegboard hooks and accessories

HDF Board: High Density Fiberboard ¹/₄" commercial-grade pegboard is a cost-effective, decorative, premium solution when a sturdier pegboard is required. The ideal organization and storage solution for garage, shed, laundry, utility room, craft room, bedroom and closet areas. Unlike regular pegboards that are constructed of layers of glued cardboard, these commercial-grade pegboards will not dry-out, warp, crack or fray - even under severe fluctuations in temperature and humidity, completely maintaining the integrity of the finish. HDF is a wood product similar to MDF, but much denser, stronger and harder. It is recommended to be used with our DuraHook® 70000 series locking pegboard hooks and accessories.

Ordering information: Limited lifetime warranty.

Delivery information: Mounting hardware not included.

Caution: Drilling, sawing, sanding, or machining wood products can expose you to wood dust, a substance known to the State of California to cause cancer. Avoid inhaling wood dust or use a dust mask or other safeguards for personal protection. For more information go to https://www.p65warnings.ca.gov/fact-sheets/wood-dust.

Order Now
 

ORDER

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty