Would you like to log in again?
Axygen® HybridRack Pipet Tips
Supplier: Corning
Axygen®
ERF-100-R-S ERF-10XT-L-R-S ERF-20-R-S ERF-10XT-R-S ER-200-R-S ERF-100-L-R-S ERF-1000XT-R-S ERF-20-L-R-S ER-300NX-R-S ER-1000XT-R-S ERF-200NX-R-S ERF-300XT-L-R-S ERF-1000XT-L-R-S ER-300XT-R-S ERF-300XT-R-S ERF-200NX-L-R-S ER-10XT-R-S
77269-868CS 422.19 USD InStock
77269-868 77269-846 77269-858 77269-866 77269-878 77269-856 77269-848 77269-860 77269-850 77269-872 77269-870 77269-864 77269-876 77269-854 77269-862 77269-852 77269-874
Axygen® HybridRack Pipet Tips
Pipette Tips
Filtered, maxymum recovery, and sterile pipet tips in a truly sustainable packaging format that reduces plastic by 70% per case.
  • Patent pending design uses 70% less plastic than traditional tip rack, equal to six pounds of plastic saved per case
  • Paperboard base is easy to recycle with most local recycling programs
  • Made in the USA in an EngergyStar, ISO 14001, and ISO 13485 certified facility
  • Combination of the strong plastic deck and sturdy paperboard base allow for pipetting directly from the HybridRack, even with a mulit-channel instrument
  • Color coded decks depending on tip volume
The HybridRack packaging format exchanges the plastic base of the pipet tip rack with a high quality, fully recyclable, paperboard base. It utilizes the polypropylene deck from our established and trusted Axygen MultiRack and Corning DeckWorks product lines in conjunction with the new paperboard base to create a stable, sustainable pipetting platform. End users can pipet directly from the HybridRack with a single or multi-channel pipettor, finish a rack of tips and simply recycle the HybridRack in their curbside recycling bin.
Using HybridRack, significantly less waste is generated from pipet tips racks: Compared to traditional plastic pipet tip racks, each case of 200 µl tips in the HybridRack packaging format saves six pounds of plastic. For 1000 µl tips, eight pounds of plastic is saved! With HybridRack, Corning takes the effort out of being sustainable so customers can focus on what really matters: their research.
Certifications: ISO 13485, ISO 14001, EnergyStar.
Packaging: Packaged in Axygen's newly redesigned sustainable pack box that uses less ink, printed on 100% recycled corrugate with an aqueous coating.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty