Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Drying and Heating Chambers, with Forced Convection, M Avantgarde.Line

Supplier: Binder
9010-0371 9010-0374 9010-0373 9010-0372
77269-970EA 9044.24 USD InStock
77269-970 77269-984 77269-976 77269-988
Drying and Heating Chambers, with Forced Convection, M Avantgarde.Line
Testing Chambers Temperature Test Chambers
The M Series Avantgarde.Line units have a maximum of 300 °C and various advanced programming options ensure these material test chambers are ideal for material and aging tests. The units have a high air flow and their powerful fans ensure rapid heating.

  • Temperature range: 15 to 22 °C above ambient temperature up to +300 °C
  • High temperature accuracy thanks to APT.line™ technology with increased heating output
  • Reinforced fan with adjustable speed
  • Electromechanical control of the exhaust air flap
  • Intuitive touchscreen controller with time-segment and real-time programming
  • Class 2 independent adjustable temperature safety device (DIN 12880) and class 3.1 independent temperature safety device (DIN 12880) with visual/acoustic alarm available for selection on the controller
  • Failsafe function - backup sensor PT100
  • Computer interface: Ethernet

Units have an internal data logger, the measured values can be read out in open format via USB.

Heat-up time: 10 min to 150 °C (M 56), 11 minutes M115, 13 minutes M 260 and 25 minutes M 720.

Certifications: IP protection class according to DIN EN 60529: IP 20.

Delivery information: Supplied with two chrome plated racks.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty