Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Code Corporation Code Reader™ 5200 Barcode Scanner

Supplier: Brady
Code Corporation
176696 176695
77282-158EA 575.28 USD InStock
77282-158 77282-160
Code Corporation Code Reader™ 5200 Barcode Scanner
Barcode Systems
Seamlessly connect your operations with the CR5200. It reads barcodes in any orientation, plus offers hands-free operation to expedite workflows.

  • Rotational tolerance ±180°
  • Field of view 51° horizontal by 39.4° vertical
  • CMOS 1.2 megapixel sensor
  • Dark gray color
  • Cabled scanner connectivity (6' cable length)
  • No battery

Get high-speed, omnidirectional reading of 1D and 2D barcode symbologies. Quickly read barcodes on mobile device screens. Compact and adjustable for any reading direction. Provides user feedback with LED and audible tone. Features extended read ranges.

Model 176696 features age verification when scanning ID cards.

Delivery information: Includes (1) CR5200; (1) USB cable.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty