Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Code Corporation Code Reader™ 1100 Barcode Scanner

Supplier: Brady
Code Corporation
176698 176697
77282-162EA 473.76 USD InStock
77282-162 77282-164
Code Corporation Code Reader™ 1100 Barcode Scanner
Barcode Systems
Automate your operations with the CR1100. With industrial-grade durability and consumer-grade functionality, you get a seamless experience with every scan.

  • Select high density or wide field
  • Rotational tolerance ±180°
  • Field of view: high density field: 30° horizontal by 20° vertical; wide field: 50° horizontal by 33.5° vertical
  • CMOS 1.2 megapixel sensor
  • Dark gray color
  • Cabled scanner connectivity (6' cable length)
  • No battery

Read more types of barcodes with patented dual-field optics. Get extra protection to seal out dust and moisture with an IP 54 rating. Lightweight and compact with rugged design. Is customizable for workflow needs with visual, audible and haptic indicators. Features a powerful JavaScript platform for complete device control.

Model 176698 includes a stand to protect scanner while not in use.

Delivery information: Includes (1) CR1100; (1) USB cable. Available with or without optional stand.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty