Would you like to log in again?
Industrial Label Printer with Brady Workstation software Barcode Scanner Kit, i7100
Supplier: Brady
176706 176714 176722
77282-212KT 6646.18 USD InStock
77282-212 77282-180 77282-196
Industrial Label Printer with Brady Workstation software Barcode Scanner Kit, i7100
Printers Label Printers
Optimize your operations with a Brady printer and scanner system. Get a printer, scanner and software kit - advanced tools that make workflows more seamless and boost productivity.
  • Single color printing capability
  • Printer connectivity options: Ethernet; RS232; USB 2.0; USB host
  • No printer cutter
  • PC connected and stand-alone
  • Scanner field of view: high density field: 30° horizontal by 20° vertical; wide field: 50° horizontal by 33.5° vertical
i7100 label printer - Print ultra-sharp labels with this 600 dpi printer - as small as 0.125" Printer is designed to print thousands of labels per day with high accuracy. Create labels with text or graphics with the included label creation software.
CR950 Scanner - Get high-speed omnidirectional reading of 1D and 2D barcodes. Scanner offers glare reduction technology for reading barcodes on shiny surfaces. The CR950 provides easier scanning with a wide area image sensor.
CR1500 Scanner - Omnidirectional scanner eliminates repositioning and rescans of 1D and 2D barcodes. Scanner is compact, lightweight and rugged to survive your toughest industrial environments. Read both high-density and wide-field barcodes with a device via dual-field optics.
CR2700 Scanner - Easily read wide and small barcodes with the ultra-fast CR2700 handled barcode scanner. Omnidirectional scanner eliminates repositioning and time-consuming re-scans of barcodes. The CR2700 provides unequaled performance reading on curved and shiny surfaces. Scanner offers customizable visual, audible and haptic feedback with multiple programmable buttons.
Delivery information: Model CR950 includes (1) Brady workstation product and wire ID software suite (BWS-PWID-EM); (1) BradyPrinter i7100 600 dpi printer (149056); (1) Configuration Settings Manual; (1) Owners Manual; (1) Product CD with Driver; (1) U.S. Type B Plug Power cord; (1) USB cable; (2) 1 In Ribbon Takeup Cores; (1) CR950 Handheld wired barcode scanner (176522).
Model CR1500 includes (1) Brady workstation product and wire ID software suite (BWS-PWID-EM); (1) BradyPrinter i7100 600 dpi printer (149056); (1) Configuration Settings Manual; (1) Owners Manual; (1) Product CD with Driver; (1) U.S. Type B Plug Power cord; (1) USB cable; (2) 1 In Ribbon Takeup Cores; (1) CR1500 Handheld wired barcode scanner with stand (176511).
Model CR2700 includes (1) Brady workstation product and wire ID software suite (BWS-PWID-EM); (1) BradyPrinter i7100 600 dpi printer (149056); (1) Configuration Settings Manual; (1) Owners Manual; (1) Product CD with Driver; (1) U.S. Type B Plug Power cord; (1) USB cable; (2) 1 In Ribbon Takeup Cores; (1) CR2700 Scanner handled scanner with charging station (176516).
Caution: CR2700 scanner has lithium-ion battery - must ship ground.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty