Would you like to log in again?
BCR® Heavy Weight Socks
Supplier: Berkshire
BCR®
BCRSOCKH3014B
77282-226CS 1595 USD InStock
77282-226
BCR® Heavy Weight Socks
Workwear
BCR® Heavy Weight socks are made from 99% continuous filament polyester and 1% polyester-covered spandex. These thick and lofty yarns provide a sock engineered for comfort
even when worn with rigid safety shoes or for extended periods.
  • Constructed using a special knitting technique; flat toe seams minimize abrasion, even when worn with steel-toe shoes
  • One size fits all
  • Designed and manufactured principally for cleanroom applications
  • Laundered and packaged in an ISO Class 4 cleanroom
  • No-run and non-ravel
  • Reduced odor caused by foot perspiration
  • Flat toe seams and stretchy elastic cuffs for operator comfort
The BCR® Heavy Weight Cleanroom socks have a flat toe seam which prevents annoying chafing during long wear and the stretchy elastic cuff ensures the sock stays in place without constricting the calf. BCR® Heavy Weight Socks are a tubular one-size-fits-all style and designed with a mid-calf length.
Autoclavable: 121 °C, 30 min.
Certifications: Meets the requirements of EU GMP Annex 1 section 7.14.
Packaging: 30 pairs/pack, 14 packs/case, bulk packaged case of 420 pairs/840 individual.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty