Would you like to log in again?
DualClean™ Softwall Strip Cleaner, Contec
Supplier: Contec
DCQT0001 P191061E DCQT0002 2746S 2778 2778 DCMD0001 DCMD0001
10797-032EA 419.4 USD InStock
10797-032 10797-034 CA10797-032 10797-036 77282-228 10797-038 76121-376 CA10797-038
DualClean™ Softwall Strip Cleaner, Contec
Mops
The DualClean™ Softwall Strip Cleaner is ideal for cleaning strip curtains and barriers in clean workspace enclosures and ISO Class 5 isolated cleanroom work cells.
  • Stainless steel frame securely holds wiping covers for uniform strip contact
  • Cleans both sides of strip curtain, up to 12" wide in one pass
  • Tension on strip is easily controlled
  • Prevents cleaning covers from twisting and binding
  • Can be sterilized by most common methods, making it ideal for critical environments
  • Traps and removes contaminants
  • Extremely clean and durable
  • Compatible with most common solvents and disinfectants
The DualClean™ Softwall Strip Cleaner is a unique system that enables one operator to easily and quickly clean both sides plastic strip curtains in a single pass, up to 12" in width. The system consists of a fabric cover that is snugly installed over a stainless steel frame with two curved-surface arms. Each curtain strip to be cleaned is positioned between the two arms of the frame, with the arms contacting both sides of the curtain at once. With slight tension maintained on the curtain strip, the cleaner is moved down the length of the strip, easily and uniformly cleaning both sides of the entire length of the strip.
The DualClean™ system can be used with two different fabric covers to provide the best cleaning performance for a variety of curtain types and environments. The microfiber cover removes and traps dust and other visible contaminants. The Quiltec® knit polyester cover is ideally suited to more critical environments. For highly critical environments like ISO Class 5 cleanrooms, the Quiltec® cover is available sterile and triple bagged. Using the QuickConnect™ system, the DualClean™ frame may be attached to a wide variety of handles. The short telescoping fiberglass handle is recommended. The electropolished, stainless steel frame is resistant to most chemicals and disinfectants and may be sterilized by common methods including gamma irradiation, ethylene oxide, or autoclave.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty