Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Code Corporation Code Reader™ 950 Barcode Scanner with USB Cable

Supplier: Brady
Code Corporation
176836
77282-346EA 284.26 USD InStock
77282-346
Code Corporation Code Reader™ 950 Barcode Scanner with USB Cable
Barcode Systems
Improve data collection operations with this swift and accurate barcode reader. The CR950 thrives in fast-paced environments, offering increased efficiency with its intuitive targeting device.

  • Rotational tolerance: ±180°
  • Field of view: 51° horizontal by 39.4° vertical
  • CMOS 1.2 megapixel monochrome sensor
  • High density field selection
  • Black color
  • 6' cable length

Get high speed omnidirectional reading of 1D and 2D barcodes and glare reduction technology for reading barcodes on shiny surfaces. Offers manual or automatic triggering. Provides easier scanning with a wide area image sensor. Features two programmable indicators: LED and tone. No battery.

Add model 176857 durable stand for hands-free scanning or for securing your scanner when not in use.

Delivery information: Includes a 1.8 m (6') USB cable and (1) CR950 reader.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty