Would you like to log in again?
Code Corporation CR2700 Barcode Scanner Accessories
Supplier: Brady
Code Corporation
176854 176508 176855 176853 176699
77282-156EA 33.84 USD InStock
77282-156 77282-354 77282-166 77282-356 77282-358
Code Corporation CR2700 Barcode Scanner Accessories
Barcode Systems
Accessories available for the Barcode Scanner CR2700.
Cable (176508) - Use this 6-foot straight cable for making USB to Micro USB connections. Reader termination: Micro USB. Host termination: USB Type A.
B27 lithium-ion battery (176699) - powers the CR2700 reader with a unique set of features that keep workflow interruptions to a minimum. Power gauge to track battery level. Quick-release latch allows for toolless swapping. The B27 battery can charge while installed in a CR2700 reader using the inductive charging station (CRA-A271).
Quad bay charging station (176855) - Keep your CR2700 batteries charged with this 4-bay charging dock. Includes charging dock and power cord (international power supply). For use with a CR2700 B27 battery.
Charging station (176853 and 176854) keep your CR2700 charged with this inductive charging station. Inductive charging eliminates exposed metal and related contact failure caused by chemical wipe-downs. Features a base and a 0.91 m (3') USB cable (international power supply).
Caution: Model 176699 has lithium-ion battery - must ship ground.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty