Would you like to log in again?
24 Hour Urine Containers
Supplier: Globe Scientific
108010 108030A 108020A 108025A
77296-328CS 236.73 USD InStock
77296-328 77296-326 77296-324 77296-322
24 Hour Urine Containers
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
These 24-hour urine collection containers offer sizes, configurations and features required to meet most 24-hour collection needs.
  • Amber polyethylene (PE) construction
  • Easy to read molded graduations
  • Molded handles for easy handling
  • Leak resistant closures
  • TransferTop™ option for improved laboratory safety and hygiene
Upright configurations for efficient storage and low form designs for stability, these are all designed with easy to read, molded graduations and leak resistant closures. Amber polyethylene construction protects samples from light and molded handles make handling easy. The new TranferTop™ option includes an integrated mechanism for 'closed system' transfer to vacuum urine sample tubes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty