Would you like to log in again?
Diamond® RealSeal™ PETG Media Bottles
Supplier: Globe Scientific
Diamond® RealSeal™
7610250 7610060 7612000 7610125 7610500 7610030 7611000
77308-586CS 223.2 USD InStock
77308-586 77308-596 77308-588 77308-598 77308-592 77308-594 77308-590
Diamond® RealSeal™ PETG Media Bottles
Bottles Plastic Bottles
Diamond® RealSeal™ PETG Square Media Bottles are an ideal solution for protecting media, laboratory reagents, and buffers during storage and transport.
  • ISO 9001 and ISO 13485 production facilty produced and packaged in ISO Class 8 cleanroom
  • Medical grade resins to meet demanding requirements
  • Leakproof, linerless caps for secure closing
  • Gamma sterilized
  • PETG bottle material protects contents during storage
  • Limits transmission of O₂ and CO₂ to protect pH sensitive solutions
Diamond® RealSeal™ PETG Square Media Bottles protect media, laboratory reagents, and buffers during storage and transport. Perfect for cold storage, the PETG bottles remain shatterproof even at −40 °C. Low transmission rates of O₂ and CO₂ maintain optimum pH values for solutions in storage. Bottles include white HDPE caps with integrated seal rings and matching semi-buttressed threads for leakproof performance to 95kPa. Bottles are provided sterile and ready to fill with tamper-evident shrink bands in tray packs for convenience.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty