Would you like to log in again?
Epredia™ 210 Laser Biopsy Cassettes with Front Tab and Separate Lid
Supplier: Richard-Allan Scientific
Epredia™
210-09-10LM 210-11-10LM 210-03-10LM 210-12-10LM 210-06-10LM 210-08-10LM 210-01-10LM 210-10-10LM 210-04-10LM 210-05-10LM 210-02-10LM 210-07-10LM
77308-612CS 504.5 USD InStock
77308-612 77308-622 77308-614 77308-624 77308-630 77308-632 77308-610 77308-620 77308-616 77308-626 77308-618 77308-628
Epredia™ 210 Laser Biopsy Cassettes with Front Tab and Separate Lid
Embedding Cassettes Cassettes for Labeling Instruments
Manufactured specifically for laser printing, the series 210 biopsy cassettes comes with separate lids with a tab in the front for easy opening and closing.
  • Series 210 laser cassettes are designed for use on both the Epredia™ NOVA™ and VEGA™ laser printers
  • They contain a special constituent chemical compound that carbonizes when exposed to the laser beam
  • Comes in 12 different colors: white, yellow, blue, pink, green, orange, gray, aqua, lilac, beige, ochre and red with color-matched lids
  • Taped stacks of 40 cassettes and 25 stacks per case for a total of 1000 cassettes and lids
  • Series 210 laser cassette is not compatible with thermal tape style printers
Designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into the Epredia™ laser cassette hoppers for printing.
Delivery information: Lids included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty