Would you like to log in again?
Epredia™ 615 Laser Bionet Cassettes with Separate Lids
Supplier: Richard-Allan Scientific
Epredia™
615-02-10LMI 615-04-10LMI 615-05-10LMI 615-10-10LMI 615-12-10LMI 615-01-10LMI 615-08-10LMI 615-07-10LMI 615-11-10LMI 615-03-10LMI 615-06-10LMI 615-09-10LMI
76614-874CS 605.15 USD InStock
76614-874 77308-644 77308-646 76614-872 76614-868 77308-652 77308-654 76614-876 77308-642 77308-650 76614-870 77308-648
Epredia™ 615 Laser Bionet Cassettes with Separate Lids
Embedding Cassettes Cassettes for Labeling Instruments
Manufactured specifically for laser printing, the series 515 Bionet biopsy cassettes come with separate lids and are available in 12 colors.
  • Series 615 laser cassettes are designed for use on both the Epredia™ NOVA™ and VEGA™ laser printers
  • They contain a special constituent chemical compound that carbonizes when exposed to the laser beam
  • Comes in 12 different colors: white, yellow, blue, pink, green, orange, gray, aqua, lilac, beige, ochre and red with color-matched lids
  • Taped stacks of 40 cassettes; 25 stacks per case for a total of 1000 cassettes and lids
  • Series 615 Laser Cassette is not compatible with thermal tape style printers
Designed to maximize fluid exchange during processing and come in stacks held together with string for ease of loading into the Epedia™ laser cassette hoppers for printing.
Ordering information: Lids included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty