Would you like to log in again?
Epredia™ 815 Laser Slotted Cassettes with Attached Lid
Supplier: Richard-Allan Scientific
Epredia™
815-03-18LM 815-06-18LM 815-12-18LM 815-11-18LM 815-09-18LM 815-05-18LM 815-07-18LM 815-10-18LM 815-02-18LM 815-01-18LM 815-08-18LM 815-04-18LM
76614-886CS 1210.28 USD InStock
76614-886 77308-656 76614-896 77308-658 76614-884 76614-894 76614-878 76614-888 76614-882 76614-892 76614-880 76614-890
Epredia™ 815 Laser Slotted Cassettes with Attached Lid
Embedding Cassettes Cassettes for Labeling Instruments
Manufactured specifically for laser printing, the series 815 slotted cassettes come with attached lids and are available in 12 colors.
  • Series 815 laser cassettes are designed for use on both the Epredia™ NOVA™ and VEGA™ laser printers
  • They contain a special constituent chemical compound that carbonizes when exposed to the laser beam
  • Comes in 12 different colors: white, yellow, blue, pink, green, orange, gray, aqua, lilac, beige, ochre and red with color-matched lids attached to the cassette
  • Taped stacks of 40 cassettes and 45 stacks per case for a total of 1800 cassettes and lids
  • Series 815 laser cassette is not compatible with thermal tape style printers
Designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into the Epredia™ laser cassette hoppers for printing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty