Would you like to log in again?
Epredia™ SlideMate™ Laser Slides and SlideMate™ Plus Adhesion Laser Slides
Supplier: Richard-Allan Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Epredia™ SlideMate™ Epredia™ SlideMate™ Plus
LS-8041I-8524-1 LS-4041I-PS8523-1 LS-5041I-8524-1 LS-6041I-PS8523-1 LS-4041I-8524-1 LS-7041I-8524-1 LS-6041I-8524-1 LS-7041I-PS8523-1 LS-8041I-PS8523-1 LS-5041I-PS8523-1
77320-778CS 673.69 USD InStock
77320-778 77320-776 77320-788 77320-774 77320-786 77320-772 77320-784 77320-782 77320-790 77320-780
Epredia™ SlideMate™ Laser Slides and SlideMate™ Plus Adhesion Laser Slides
Slides Microscope Slides
Optimized for use on the SlideMate™ Laser Printer, these slides provide for a clear and robust printing surface. The adhesion properties of SlideMate™ Laser Plus slides are the same technology as Superfrost PLUS™ slides.
  • The adhesive properties of the SlideMate™ Laser Plus Slides makes them suitable for all routine clinical applications
  • The tab is impervious to virtually all common laboratory chemicals and reagents
Epredia™ SlideMate™ laser slides are made from extra-white glass, manufactured to high quality standards. The clarity achieved during the manufacturing process helps ensure high levels of transparency to aid in accurate diagnoses.
The slides come pre-cleaned, flat with finely ground edges. Each package comes wrapped to help protect against humidity and moisture. The packaging is also designed to assist with loading the slides into SlideMate™ laser printer hoppers to prevent contamination due to handling. Manufactured in the USA, SlideMate™ laser slides are smooth, three-quarter inch white, blue, green, pink or yellow tab slides validated to work with the SlideMate™ laser slide printer.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty