Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Stone Mills, 230, MorehouseCowles

Supplier: MorehouseCowles

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

491159 487274
77320-858EA 29454.55 USD InStock
77320-858 77320-860
Stone Mills, 230, MorehouseCowles
Mills
Since 1925, MorehouseCowles stone mills have proven to be the most reliable and efficient way to process hundreds of different materials from food, cosmetics, and ceramics to paints, inks and grease.

  • All parts in contact with product are 316 stainless steel, except for aluminum oxide stones
  • All non-wetted parts are 304 stainless steel construction
  • 304 stainless steel base with mill finish
  • Choice of standard or Explosion proof units
  • Precision adjustment wheel permits precise stone clearance adjustment within 0.001" for consistent processing
  • Dual-bearing spindle allows positive, stable alignment of stones, connected to motor with flexible coupling for longer bearing life
  • Collet lock maintains exact adjustment, minimizes vibration
  • Quiet high-speed operation
  • Easy-to-clean process chamber

Stone mills break down a product or mixture to very small particle sizes. They offer grinding technology for dry to liquid processing where 100% of the material passes through the milling surfaces for superior consistency and performance.

MorehouseCowles stone mills feature a very simple design and process with extra heavy-duty construction that has earned them a reputation as the 'reliable workhorse'. This long-lasting reliability is only possible by using the finest materials and construction techniques. The highly polished inner surfaces facilitate easy cleaning in sanitary applications and fast product or color changes.

Spindle: 316 stainless steel
Spindle speed: 3600 rpm
Stones: 2" diameter, 54, 60, 80 and 100 grit
Product feed: Funnel or optional NPT threaded inlet or Triclover adaptor
Utilities required: Electrical power, 1 phase

Packaging: One wooden box. Dimensions (L×W×H): 20×20×37", Weight: 155.0 lbs.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty