Would you like to log in again?
Automated Roll Heat Sealer, Azenta Life Sciences
Supplier: Azenta Life Sciences
4TI-0665-57 4TI-0665-58 4TI-0665-60 4TI-0665 4TI-0665-56 4TI-0665-59
77321-766EA 217.8 USD InStock
77321-766 77321-768 77321-774 77321-770 76638-838 77321-772
Automated Roll Heat Sealer, Azenta Life Sciences
Heat Sealers Microplate Heat Sealers
The perfect solution for automated heat sealing of microplates, PCR plates, assay plates and storage plates used in medium to high-throughput or full automation applications. No separate air supply required.
  • SiLA compatible
  • Compatible with a wide range of plates and seals
  • 2 positions for different roll sizes
  • Variable time and temperature controls
  • Seal cycle time < 15 seconds
  • Color touch screen with intuitive user interface
  • Auto standby mode
The Automated Roll Heat Sealer is compatible with a wide range of SBS footprint plates, including all PCR plate formats from 96 to 1536 well, assay plates, deep well storage plates and microplates. Varying plate heights, from 3 to 60 mm can be accommodated with minimal adjustments of the instrument. Plates are presented in portrait format and the plate position on the shuttle allows good access for robotic integration. Where flexibility in applications utilizing different plate types is required in automation platforms, lightweight adaptors are available which allow the robotics to interchange the adapter.
Rolls with a width of between 75 to 85 mm can be loaded in either of two roll positions, depending on roll length and available space, enabling the entire instrument to fit under the gantry of most liquid handling robots. The width and length of the applied seals are set to precisely cover an SBS footprint plate without interfering with plate stacking. The seal position can be adjusted to give an overhang at one end of the plate to facilitate ease of seal removal.
Easy integration into robotic systems through plug and play. High-performance heating block design. Flexible with a wide range of consumables including gas permeable seals without instrument modification. It used powerful electric motor drives plate sealing mechanism for enables optimization of any seal/plate type. Unlimited password protected protocols. Rapid heating, no delay between seals, uniform temperature across entire heating block, +/- 1 degree edge-to-edge, corner-to-corner.
Certifications: CE marked.
Ordering information: Two-year warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty