Would you like to log in again?
Tyvek Sleeve Cover, Sterile
Supplier: Keystone Cleanroom Products
1804TW28CFDPST1
77321-790CS 2940.3 USD InStock
77321-790
Tyvek Sleeve Cover, Sterile
Disposable Garments
Extra-long sterile Tyvek sleeves are packaged in multiple layers of Tyvek peel pouches and poly bags to gain access into the aseptic area without excessive spraying and wiping with sanitization agents. The sleeves are individually packed and specially folded for easy aseptic donning.
  • Manufactured and packaged in an ISO certified cleanroom
  • Clean and lint free
  • Tear and puncture resistant
  • Provides an excellent barrier to particles and microorganisms
  • Heat-sealed seams eliminate particles and liquids from passing through the seams
  • Each sleeve has elastic at the arm and wrist
  • Features a special cuff fold design, for easy aseptic donning
  • Individually double packaged in Tyvek/film chevron peel pouches and then inside a third, sealed poly bag layer
  • Labeled with lot number, date of manufacture and expiration date
  • Validated sterile to a 10⁻⁶ SAL
  • Certificates of conformance and sterility sent with each shipment
Keystone cleanroom products’ extra long sterile Tyvek® sleeves provide the optimal aseptic packaging and presentation that is required by the most critical pharmaceutical processes.
Certifications: CoC and certificate of sterility.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty