Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ESD Cleanroom Washable Shoe Covers

Supplier: Transforming Technologies

SC50BSWH04 SC50BSLB09 SC50BSWH07 SC50BSWH08 SC50BSLB03 SC50BSLB08 SC50BSWH06 SC50BSLB04 SC50BSLB06 SC50BSLB05 SC50BSLB01 SC50BSWH01 SC50BSWH09 SC50BSWH02 SC50BSLB02 SC50BSWH05 SC50BSWH03 SC50BSLB07
77321-906PR 44.62 USD InStock
77321-906 77321-918 77321-908 77321-902 77321-914 77321-904 77321-916 77321-920 77321-898 77321-888 77321-910 77321-922 77321-900 77321-912 77321-894 77321-896 77321-890 77321-892
ESD Cleanroom Washable Shoe Covers
Workwear
ESD-safe integrated cleanroom wear designed for use in controlled environments.

  • TX4000 ESD grid fabric: 99% polyester/1% carbon
  • 3-layer dissipative rubber soft sole
  • Adjustable heel snap
  • Conductive ribbon connected to sole with internal snap
  • Dissipative sole and ESD canvas fender: <3.5×10e7
  • Replaceable 10"" conductive ribbon for contact with the body
  • ISO Cleanroom: ISO 6 (Class 1000)
  • Launderable

ESD washable cleanroom shoe covers with ESD rubber soles and conductive grounding ribbons are an economical way to maintain cleanliness and ESD protection in controlled environments and static sensitive areas. Ideal for cleanrooms, laboratories, manufacturing facilities, and other sterile environments that are concerned with static and ESD damage. These garments provide a path-to-ground for static charges and convert non-ESD footwear into a grounding solution.

Certifications: In compliance with ANSI/ESD S20.20 via test method ANSI/ESD STM9.1.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty